siRNA Details
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022589.1 | NP_072111.1 | 5 | 410 | ||
GenBank mRNA | AJ271442.1 | - | 5 | 410 | ||
BC085733.1 | - | 5 | 430 | 7122828 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022589.1 | NP_072111.1 | 5 | 410 | ||
GenBank mRNA | AJ271442.1 | - | 5 | 410 | ||
BC085733.1 | - | 5 | 430 | 7122828 |