siRNA Details
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 1032 | ||
NM_001270562.1 | NP_001257491.1 | 4 | 930 | |||
NM_001270563.1 | NP_001257492.1 | 4 | 890 | |||
NM_022593.4 | NP_072115.1 | 4 | 926 | |||
XM_006237724.2 | XP_006237786.1 | 4 | 927 | |||
GenBank mRNA | BC060566.1 | - | 4 | 902 | 6918038 | |
FQ213660.1 | - | 4 | 939 | |||
FQ214805.1 | - | 4 | 1032 |

More Information
Other Species Targeted

Gene Ontology Categories:
