siRNA Details
Gene Symbol: Tceb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270561.1 | NP_001257490.1 | 4 | 671 | ||
NM_001270562.1 | NP_001257491.1 | 4 | 569 | |||
NM_001270563.1 | NP_001257492.1 | 4 | 529 | |||
NM_022593.4 | NP_072115.1 | 4 | 565 | |||
XM_006237724.2 | XP_006237786.1 | 4 | 566 | |||
GenBank mRNA | BC060566.1 | - | 4 | 540 | 6918038 | |
CK355540.1 | - | 4 | 565 | |||
FM093546.1 | - | 4 | 531 | |||
FM101371.1 | - | 4 | 519 | |||
FM110992.1 | - | 4 | 535 | |||
FQ213660.1 | - | 4 | 579 | |||
FQ214805.1 | - | 4 | 671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
