siRNA Details
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022602.1 | NP_072124.1 | 783 | |||
GenBank mRNA | AF057026.1 | - | 783 | |||
AF086624.1 | - | 985 | ||||
BC097317.1 | - | 762 | 7446368 |

More Information
Other Species Targeted

Gene Ontology Categories:
