siRNA Details
Gene Symbol: Fgfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022603.1 | NP_072125.1 | 1 | 379 | ||
XM_006251078.2 | XP_006251140.1 | 2 | 513 | |||
XM_006251079.2 | XP_006251141.1 | 2 | 435 | |||
GenBank mRNA | AF142758.1 | - | 1 | 379 |
