siRNA Details
Gene Symbol: Esm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022604.2 | NP_072126.1 | 1 | 229 | ||
GenBank mRNA | BC070888.1 | - | 1 | 229 | 7098029 | |
U80818.1 | - | 1 | 142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
