siRNA Details
Gene Symbol: Ilkap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022606.1 | NP_072128.1 | 6 | 548 | ||
XM_008767296.1 | XP_008765518.1 | 5 | 2504 | |||
XM_008767297.1 | XP_008765519.1 | 4 | 2829 | |||
GenBank mRNA | AF095927.1 | - | 6 | 548 | ||
BC062010.1 | - | 6 | 550 | 5621497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
