siRNA Details
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022607.2 | NP_072129.2 | 3 | 769 | ||
GenBank mRNA | AB000098.1 | - | 3 | 700 | ||
CB608313.1 | - | 2 | 535 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
