siRNA Details
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022618.1 | NP_072140.1 | 13 | 5685 | ||
GenBank mRNA | AF139518.1 | - | 13 | 5685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
