siRNA Details
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022627.2 | NP_072149.1 | 5 | 659 | ||
XM_006233011.2 | XP_006233073.1 | 6 | 860 | |||
XM_006233012.2 | XP_006233074.1 | 6 | 760 | |||
GenBank mRNA | AF182717.1 | - | 5 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
