siRNA Details
Gene Symbol: Wnt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022631.1 | NP_072153.1 | 4 | 891 | ||
XM_006252664.2 | XP_006252726.1 | 4 | 761 | |||
XM_006252665.2 | XP_006252727.1 | 4 | 723 | |||
XM_008771007.1 | XP_008769229.1 | 4 | 1599 | |||
GenBank mRNA | AB244721.1 | - | 3 | 657 | ||
AF188333.1 | - | 4 | 891 | |||
AY819646.1 | - | 3 | 657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
