siRNA Details
Gene Symbol: Nat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022635.1 | NP_072157.1 | 1 | 612 | ||
XM_006236789.2 | XP_006236851.1 | 2 | 694 | |||
XM_006236790.2 | XP_006236852.1 | 2 | 704 | |||
GenBank mRNA | AF185570.1 | - | 1 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
