siRNA Details
Gene Symbol: Rem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022685.2 | NP_073176.2 | 3 | 556 | ||
GenBank mRNA | AF084464.1 | - | 3 | 430 | ||
AY916790.1 | - | 3 | 496 | |||
BC127495.1 | - | 3 | 541 | 8371030 |

More Information
Other Species Targeted

Gene Ontology Categories:
