siRNA Details
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022694.2 | NP_073185.2 | 3 | 485 | ||
GenBank mRNA | AY697864.1 | - | 3 | 495 | ||
BC072471.1 | - | 3 | 485 | 7100501 | ||
U83883.1 | - | 3 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
