siRNA Details
Gene Symbol: Bad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022698.1 | NP_073189.1 | 3 | 456 | ||
XM_006230896.2 | XP_006230958.1 | 3 | 1161 | |||
XM_006230897.2 | XP_006230959.1 | 2 | 339 | |||
XM_006230898.2 | XP_006230960.1 | 3 | 354 | |||
GenBank mRNA | AF031227.1 | - | 3 | 494 | ||
AF279910.1 | - | 3 | 456 | |||
AF279911.1 | - | 3 | 456 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
