siRNA Details
Gene Symbol: Mbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022704.2 | NP_073195.2 | 3 | 320 | ||
XM_006231249.2 | XP_006231311.1 | 5 | 596 | |||
XM_006231250.2 | XP_006231312.1 | 5 | 516 | |||
GenBank mRNA | AY325174.1 | - | 350 | |||
AY325178.1 | - | 350 | ||||
FQ210356.1 | - | 5 | 505 | |||
M14103.1 | - | 5 | 490 | |||
X05023.1 | - | 3 | 320 |
Gene Symbol: LOC100911854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749087.3 | XP_003749135.1 | 596 | |||
XM_003749088.3 | XP_003749136.1 | 516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
