siRNA Details
Gene Symbol: Gabarapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022706.2 | NP_073197.1 | 2, 3 | 165 | ||
GenBank mRNA | AB003515.1 | - | 2, 3 | 203 | ||
BC058145.1 | - | 2, 3 | 168 | 6921109 | ||
BC088139.1 | - | 2, 3 | 165 | 7323806 | ||
FQ216852.1 | - | 2, 3 | 179 | |||
FQ226397.1 | - | 2, 3 | 189 | |||
FQ231643.1 | - | 2, 3 | 158 | |||
FQ233551.1 | - | 2, 3 | 200 | |||
FQ234861.1 | - | 2, 3 | 158 |
Gene Symbol: LOC100359937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 154 |
