siRNA Details
Gene Symbol: Gabarapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022706.2 | NP_073197.1 | 3 | 195 | ||
GenBank mRNA | AB003515.1 | - | 3 | 233 | ||
BC058145.1 | - | 3 | 198 | 6921109 | ||
BC088139.1 | - | 3 | 195 | 7323806 | ||
FQ216852.1 | - | 3 | 209 | |||
FQ226397.1 | - | 3 | 219 | |||
FQ231643.1 | - | 3 | 188 | |||
FQ233551.1 | - | 3 | 230 | |||
FQ234861.1 | - | 3 | 188 |
Gene Symbol: LOC100359937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729947.4 | XP_002729993.3 | 1 | 184 |

More Information
Other Species Targeted

Gene Ontology Categories:
