siRNA Details
Gene Symbol: Pln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022707.2 | NP_073198.1 | 2 | 278 | ||
GenBank mRNA | CK356371.1 | - | 2 | 275 | ||
FM050525.1 | - | 2 | 278 | |||
S95849.1 | - | 2 | 256 | |||
S95853.1 | - | 1 | 98 | |||
X71068.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
