siRNA Details
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022711.4 | NP_073202.1 | 4, 5 | 753 | ||
GenBank mRNA | M95058.1 | - | 4, 5 | 753 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022711.4 | NP_073202.1 | 4, 5 | 753 | ||
GenBank mRNA | M95058.1 | - | 4, 5 | 753 |