siRNA Details
Gene Symbol: Srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022711.4 | NP_073202.1 | 4, 5 | 746 | ||
GenBank mRNA | M95058.1 | - | 4, 5 | 746 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
