siRNA Details
Gene Symbol: Crhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022714.1 | NP_073205.1 | 6 | 787 | ||
XM_006236556.2 | XP_006236618.1 | 7 | 777 | |||
XM_006236557.2 | XP_006236619.1 | 7 | 774 | |||
XM_006236558.1 | XP_006236620.1 | 6 | 805 | |||
GenBank mRNA | EF078964.1 | - | 6 | 572 | ||
U16253.1 | - | 6 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
