siRNA Details
Gene Symbol: Nucks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022799.2 | NP_073636.1 | 6 | 603 | ||
XM_006249797.2 | XP_006249859.1 | 6 | 672 | |||
GenBank mRNA | AJ237669.1 | - | 5 | 428 | ||
BC078818.1 | - | 6 | 603 | 7128528 | ||
FQ228159.1 | - | 6 | 628 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
