siRNA Details
Gene Symbol: Nab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244902.2 | XP_006244964.1 | 2 | 565 | ||
GenBank mRNA | FM034218.1 | - | 2 | 533 | ||
FQ222344.1 | - | 3 | 541 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008667, U47008, AB221677, BC005627, BC016886.
Gene Ontology Categories:
