siRNA Details
Gene Symbol: Foxq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022858.2 | NP_074049.2 | 1 | 992 | ||
GenBank mRNA | BC161861.1 | - | 1 | 992 | ||
L13201.1 | - | 1 | 630 |

More Information
Other Species Targeted

Gene Ontology Categories:
