siRNA Details
Gene Symbol: Rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022931.1 | NP_075220.1 | 5 | 879 | ||
GenBank mRNA | AF199334.1 | - | 5 | 879 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022931.1 | NP_075220.1 | 5 | 879 | ||
GenBank mRNA | AF199334.1 | - | 5 | 879 |