siRNA Details
Gene Symbol: Rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022931.1 | NP_075220.1 | 4 | 678 | ||
GenBank mRNA | AF199334.1 | - | 4 | 678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
