siRNA Details
Gene Symbol: Ephx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022936.1 | NP_075225.1 | 12 | 1170 | ||
XM_006252147.2 | XP_006252209.1 | 11 | 1159 | |||
GenBank mRNA | BC085732.1 | - | 12 | 1175 | 7188942 | |
FQ213818.1 | - | 11 | 1107 | |||
FQ216038.1 | - | 2 | 174 | |||
X65083.1 | - | 12 | 1170 |
