siRNA Details
Gene Symbol: Ephx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022936.1 | NP_075225.1 | 16 | 1430 | ||
XM_006252147.2 | XP_006252209.1 | 15 | 1419 | |||
GenBank mRNA | BC085732.1 | - | 16 | 1435 | 7188942 | |
FQ213818.1 | - | 15 | 1367 | |||
FQ213968.1 | - | 15 | 1367 | |||
FQ216038.1 | - | 6 | 434 | |||
X60328.1 | - | 2 | 42 | |||
X65083.1 | - | 16 | 1430 |
