siRNA Details
Gene Symbol: Ephx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022936.1 | NP_075225.1 | 4 | 430 | ||
XM_006252147.2 | XP_006252209.1 | 3 | 419 | |||
GenBank mRNA | BC085732.1 | - | 4 | 435 | 7188942 | |
FQ213818.1 | - | 3 | 367 | |||
FQ213968.1 | - | 3 | 367 | |||
X65083.1 | - | 4 | 430 |
