siRNA Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270838.1 | NP_001257767.1 | 9 | 1164 | ||
NM_001270839.1 | NP_001257768.1 | 9 | 1142 | |||
NM_001270840.1 | NP_001257769.1 | 9 | 1161 | |||
NM_022941.4 | NP_075230.1 | 9 | 1146 | |||
XM_006250261.2 | XP_006250323.1 | 9 | 1252 | |||
XM_008769759.1 | XP_008767981.1 | 8 | 2109 | |||
GenBank mRNA | AB104736.1 | - | 9 | 1199 | ||
AB105072.1 | - | 8 | 1290 | |||
AB204897.1 | - | 8 | 1012 | |||
AB204898.1 | - | 8 | 1015 | |||
AB204899.1 | - | 8 | 993 | |||
AB204900.1 | - | 7 | 1192 | |||
AF133094.1 | - | 8 | 997 | |||
AF133095.1 | - | 8 | 997 | |||
BC089763.1 | - | 9 | 1161 | 7324318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
