siRNA Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270839.1 | NP_001257768.1 | 8, 9 | 1096 | ||
NM_022941.4 | NP_075230.1 | 8, 9 | 1100 | |||
XM_006250261.2 | XP_006250323.1 | 8, 9 | 1206 | |||
GenBank mRNA | AB104736.1 | - | 8, 9 | 1153 | ||
AB105072.1 | - | 7, 8 | 1244 | |||
AB204899.1 | - | 7, 8 | 947 | |||
AB204900.1 | - | 6, 7 | 1146 | |||
AF133094.1 | - | 7, 8 | 951 | |||
AF133095.1 | - | 7, 8 | 951 |

More Information
Other Species Targeted

Gene Ontology Categories:
