siRNA Details
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB104736.1 | - | 8 | 1110 | ||
AB105072.1 | - | 7 | 1201 | |||
AB204897.1 | - | 7 | 905 | |||
AB204898.1 | - | 7 | 908 | |||
AB204899.1 | - | 7 | 904 | |||
AB204900.1 | - | 6 | 1103 | |||
AF133095.1 | - | 7 | 908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
