siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767428.1 | XP_008765650.1 | 6 | 1904 | ||
XM_008767429.1 | XP_008765651.1 | 6 | 1104 | |||
XM_008767430.1 | XP_008765652.1 | 6 | 1135 | |||
XM_008767431.1 | XP_008765653.1 | 6 | 1129 | |||
XM_008767432.1 | XP_008765654.1 | 6 | 1579 | |||
XM_008767433.1 | XP_008765655.1 | 6 | 791 | |||
GenBank mRNA | AB003594.1 | - | 6 | 777 | ||
FQ212368.1 | - | 6 | 893 | |||
U67137.1 | - | 8 | 1924 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001128181, NM_027712, NM_001128180, NM_177639, AK035763, and others.
Gene Ontology Categories:
