siRNA Details
Gene Symbol: Dlgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767428.1 | XP_008765650.1 | 6 | 2078 | ||
XM_008767429.1 | XP_008765651.1 | 6 | 1278 | |||
XM_008767430.1 | XP_008765652.1 | 6 | 1309 | |||
XM_008767431.1 | XP_008765653.1 | 6 | 1303 | |||
XM_008767432.1 | XP_008765654.1 | 6 | 1753 | |||
XM_008767433.1 | XP_008765655.1 | 6 | 965 | |||
GenBank mRNA | AB003594.1 | - | 6 | 951 | ||
FQ212368.1 | - | 6 | 1067 | |||
U67137.1 | - | 8 | 2098 | |||
U67987.1 | - | 6 | 892 |

More Information
Other Species Targeted

Gene Ontology Categories:
