siRNA Details
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022955.1 | NP_075244.1 | 4422 | |||
GenBank mRNA | AB011532.1 | - | 4422 |
Gene Symbol: LOC100911486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764417.1 | XP_008762639.1 | 32 | 4083 |
