siRNA Details
Gene Symbol: Cldn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031699.2 | NP_113887.2 | 2 | 493 | ||
GenBank mRNA | AF195500.1 | - | 2 | 434 | ||
BC061992.1 | - | 2 | 493 | 5599462 | ||
FQ220868.1 | - | 2 | 462 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
