siRNA Details
Gene Symbol: Cldn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031701.2 | NP_113889.1 | 1 | 731 | ||
GenBank mRNA | AF241260.1 | - | 1 | 562 | ||
BC082073.1 | - | 1 | 729 | 7133096 | ||
FQ213419.1 | - | 1 | 745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
