siRNA Details
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031702.1 | NP_113890.1 | 4, 5 | 774 | ||
XM_006246805.2 | XP_006246867.1 | 3, 4 | 974 | |||
GenBank mRNA | AJ011811.1 | - | 3, 4 | 410 | ||
BC101892.1 | - | 4, 5 | 774 | 7930797 | ||
BC159404.1 | - | 3, 4 | 868 | 7458165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
