siRNA Details
Gene Symbol: Rps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031706.1 | NP_113894.1 | 4 | 252 | ||
GenBank mRNA | BC082802.1 | - | 3 | 236 | 7302521 | |
BC166878.1 | - | 4 | 252 | 6920981 | ||
FQ221315.1 | - | 4 | 252 | |||
FQ222655.1 | - | 4 | 289 | |||
FQ222951.1 | - | 4 | 250 | |||
FQ223011.1 | - | 4 | 252 | |||
FQ223286.1 | - | 4 | 254 | |||
FQ229267.1 | - | 4 | 252 | |||
X06423.1 | - | 4 | 252 |

More Information
Other Species Targeted

Gene Ontology Categories:
