siRNA Details
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008553.1 | NP_001008553.1 | 8 | 1165 | ||
XM_006255601.2 | XP_006255663.1 | 8 | 1338 | |||
XM_008772529.1 | XP_008770751.1 | 8 | 1338 | |||
GenBank mRNA | X80778.1 | - | 8 | 1165 |
