siRNA Details
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031727.2 | NP_113915.2 | 13, 14 | 1754 | ||
XM_006249185.2 | XP_006249247.1 | 12, 13 | 4846 | |||
GenBank mRNA | CV112377.1 | - | 3, 4 | 384 | ||
D31873.1 | - | 13, 14 | 1779 |
