siRNA Details
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031736.1 | NP_113924.1 | 7 | 2213 | ||
GenBank mRNA | BC081766.1 | - | 7 | 1550 | 7106977 | |
D85100.1 | - | 7 | 2213 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc27a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031736.1 | NP_113924.1 | 7 | 2213 | ||
GenBank mRNA | BC081766.1 | - | 7 | 1550 | 7106977 | |
D85100.1 | - | 7 | 2213 |