siRNA Details
Gene Symbol: Kcnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032462.2 | NP_115851.1 | 6 | 553 | ||
XM_006234989.2 | XP_006235051.1 | 6 | 682 | |||
XM_006234990.2 | XP_006235052.1 | 5 | 3998 | |||
GenBank mRNA | AB043892.1 | - | 5 | 532 | ||
AF297118.1 | - | 6 | 555 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
