siRNA Details
Gene Symbol: Dpysl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023023.1 | NP_075412.1 | 8 | 961 | ||
GenBank mRNA | AB029432.1 | - | 7 | 835 | ||
AJ131436.1 | - | 8 | 961 |
Gene Symbol: LOC103692570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764540.1 | XP_008762762.1 | 1072 |
