siRNA Details
Gene Symbol: Dpysl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023023.1 | NP_075412.1 | 10 | 1235 | ||
GenBank mRNA | AB029432.1 | - | 9 | 1109 | ||
AJ131436.1 | - | 10 | 1235 |
Gene Symbol: LOC100911610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239841.2 | XP_006239903.1 | 2 | 805 |
Gene Symbol: LOC103692570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764540.1 | XP_008762762.1 | 1346 |
