siRNA Details
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023093.1 | NP_075581.1 | 7 | 971 | ||
GenBank mRNA | BC061830.1 | - | 7 | 971 | 5599635 | |
FQ214146.1 | - | 6 | 813 | |||
FQ225416.1 | - | 2 | 286 | |||
FQ225735.1 | - | 2 | 286 | |||
FQ227531.1 | - | 2 | 286 | |||
FQ227720.1 | - | 1 | 106 | |||
J05266.1 | - | 7 | 892 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
