siRNA Details
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023094.3 | NP_075582.3 | 5 | 320 | ||
GenBank mRNA | AB081649.1 | - | 6 | 395 | ||
AF264018.2 | - | 5 | 320 | |||
AF469945.1 | - | 5 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
