siRNA Details
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023096.2 | NP_075584.1 | 5 | 1104 | ||
XM_008764847.1 | XP_008763069.1 | 5 | 1824 | |||
GenBank mRNA | AF196965.1 | - | 5 | 915 | ||
AF385401.1 | - | 5 | 1104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
