siRNA Details
Gene Symbol: Gucy1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023956.1 | NP_076446.1 | 5 | 2060 | ||
GenBank mRNA | AB079780.1 | - | 2 | 486 | ||
AB096080.1 | - | 5 | 2060 | |||
AB097860.1 | - | 5 | 1640 | |||
AF109963.2 | - | 5 | 2060 | |||
AJ001041.1 | - | 1 | 46 |
